2-Chloro-N-methylaniline 97%

Supplier: Ambeed
Warning

A142281-10G A142281-1G A142281-25G A142281-5G
77258-868EA 63.86 USD
77258-868 77258-870 77258-872 77258-874
2-Chloro-N-methylaniline 97%
2-Chloro-N-methylaniline

Precaución: for Research Use Only

Formula: C₇H₈ClN
MW: 141.6 g/mol
MDL Number: MFCD00045170
CAS Number: 932-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR