4'-tert-Butylbiphenyl-4-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
4'-(tert-Butyl)-[1,1'-biphenyl]-4-carboxylic acid
A142273-1G
A142273-250MG
A142273-5G
77285-808EA
26.78
USD
77285-808
77285-810
77285-812
77285-814
4'-tert-Butylbiphenyl-4-carboxylic acid 95%
4'-tert-Butylbiphenyl-4-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₇H₁₈O₂ |
MDL Number:
MFCD01631300 CAS Number: 5748-42-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...