2-Fluoro-5-nitrobenzylalcohol 97%

Supplier: Ambeed
Warning

Synonyms: (2-Fluoro-5-nitrophenyl)methanol

A142266-1G A142266-250MG A142266-25G A142266-5G
77258-860EA 24.72 USD
77258-860 77258-862 77258-864 77258-866
2-Fluoro-5-nitrobenzylalcohol 97%
2-Fluoro-5-nitrobenzylalkohol

Precaución: for Research Use Only

Formula: C₇H₆FNO₃
MW: 171.1300 g/mol
MDL Number: MFCD02683502
CAS Number: 63878-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR