(2S)-1-[(R)-α-(Dimethylamino)-2-(diphenylphosphino)benzyl]-2-diphenylphosphinoferrocene 98%

Supplier: Ambeed
Warning

A142143-100MG A142143-1G A142143-250MG A142143-5G
77246-986EA 38 USD
77246-986 77246-988 77246-990 77746-004
(2S)-1-[(R)-α-(Dimethylamino)-2-(diphenylphosphino)benzyl]-2-diphenylphosphinoferrocene 98%
(2S)-1-[(R)-α-(Dimethylamino)-2-(diphenylphosphino)benzyl]-2-diphenylphosphinoferrocene

Precaución: for Research Use Only

Formula: C₄₃H₃₉FeNP₂
MDL Number: MFCD04117709
CAS Number: 1003012-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR