Cilengitide 98%

Supplier: Ambeed
Warning

Synonyms: EMD-121974, 2-((2S,5R,8S,11S)-5-Benzyl-11-(3-guanidinopropyl)-8-isopropyl-7-methyl-3,6,9,12,15-pentaoxo-1,4,7,10,13-pentaazacyclopentadecan-2-yl)acetic acid, cyclo(L-arginylglycyl-L-α-aspartyl-D-phenylalanyl-N-methyl-L-valyl)

A142071-1MG A142071-50MG A142071-100MG
77235-020EA 11 USD
77235-020 77600-060 77615-134
Cilengitide 98%
Cilengitide

Precaución: for Research Use Only

Formula: C₂₇H₄₀N₈O₇
MDL Number: MFCD04307743
CAS Number: 188968-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR