tert-Butyl 4-((2-(diethylamino)ethyl)carbamoyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate 97%

Supplier: Ambeed
Warning

A142040-1G A142040-25G A142040-5G
77258-844EA 11.33 USD
77258-844 77258-846 77258-848
tert-Butyl 4-((2-(diethylamino)ethyl)carbamoyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate 97%
tert-Butyl 4-((2-(diethylamino)ethyl)carbamoyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₃₁N₃O₃
MW: 337.45 g/mol
MDL Number: MFCD13181868
CAS Number: 590424-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR