3-Chloropyridine-2-carboxamide 95%

Supplier: Ambeed
Warning

Synonyms: 3-Chloropicolinamide

A142036-100MG A142036-1G A142036-250MG
77272-510EA 11.33 USD
77272-510 77272-512 77272-514
3-Chloropyridine-2-carboxamide 95%
3-Chloropyridine-2-carboxamide

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O
MW: 156.57 g/mol
MDL Number: MFCD10574691
CAS Number: 114080-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR