4-Benzyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-aspartate 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Asp(OBzl)-OH, 4-Benzyl N-Fmoc-L-aspartate, N-Fmoc-L-aspartic Acid 4-Benzyl Ester, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 4-Benzyl Ester

A141931-100G A141931-10G A141931-1G A141931-25G A141931-5G
77272-498EA 119 USD
77272-498 77272-500 77272-502 77272-504 77272-506
4-Benzyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-aspartate 97%
4-Benzyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-aspartate

Precaución: for Research Use Only

Formula: C₂₆H₂₃NO₆
MDL Number: MFCD00065630
CAS Number: 86060-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR