4-Hydroxy-2-methylbenzaldehyde 97%

Supplier: Ambeed
Warning

A141855-10G A141855-1G A141855-250MG A141855-5G A141855-25G
77246-980EA 92 USD
77246-980 77246-982 77246-984 77615-132 77746-002
4-Hydroxy-2-methylbenzaldehyde 97%
4-Hydroxy-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₂
MW: 136.1479 g/mol
MDL Number: MFCD04037443
CAS Number: 41438-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR