(2-Methylbenzo[d]thiazol-5-yl)methanol 97%

Supplier: Ambeed
Warning

A141821-100MG A141821-1G A141821-250MG
77235-010EA 87.55 USD
77235-010 77235-012 77235-014
(2-Methylbenzo[d]thiazol-5-yl)methanol 97%
(2-Methylbenzo[d]thiazol-5-yl)methanol

Precaución: for Research Use Only

Formula: C₉H₉NOS
MW: 179.23 g/mol
MDL Number: MFCD00843881
CAS Number: 32770-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR