2-Fluoro-6-iodobenzoic acid 98%

Supplier: Ambeed
Warning

A141474-100G A141474-10G A141474-1G A141474-25G A141474-5G
77258-810EA 188 USD
77258-810 77258-812 77258-814 77258-816 77258-818
2-Fluoro-6-iodobenzoic acid 98%
2-Fluoro-6-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄FIO₂
MDL Number: MFCD00042289
CAS Number: 111771-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR