4-Methoxysalicylaldehyde 98%

Supplier: Ambeed
Warning

A141471-100G A141471-10G A141471-1G A141471-500G A141471-5G A141471-25G A141471-1000G
77246-962EA 56 USD
77246-962 77246-964 77246-966 77246-968 77246-970 77316-164 77615-128
4-Methoxysalicylaldehyde 98%
4-Methoxysalicylaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₃
MDL Number: MFCD00003327
CAS Number: 673-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR