2,5-Dimethyl-4-hydroxy-3(2H)-furanone 98%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxy-2,5-dimethyl-3(2H)-furanone, Furaneol, 4-Hydroxy-2,5-dimethylfuran-3(2H)-one

A141458-100G A141458-10G A141458-25G A141458-5G A141458-500G
77285-756EA 107.12 USD
77285-756 77285-758 77285-760 77285-762 77747-995
2,5-Dimethyl-4-hydroxy-3(2H)-furanone 98%
2,5-Dimethyl-4-hydroxy-3(2H)-furanone

Precaución: for Research Use Only

Formula: C₆H₈O₃
MW: 128.13 g/mol
MDL Number: MFCD00010706
CAS Number: 3658-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR