2-[(4R)-4-(1,1-Dimethylethyl)-4,5-dihydro-2-oxazolyl]pyridine ≥98%, ee 99%

Supplier: Ambeed
Warning

A141321-100MG A141321-1G A141321-250MG A141321-25G A141321-5G
77246-948EA 48.41 USD
77246-948 77246-950 77246-952 77246-954 77246-956
2-[(4R)-4-(1,1-Dimethylethyl)-4,5-dihydro-2-oxazolyl]pyridine ≥98%, ee 99%
2-[(4R)-4-(1,1-Dimethylethyl)-4,5-dihydro-2-oxazolyl]pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O
MW: 204.26 g/mol
MDL Number: MFCD30541658
CAS Number: 242482-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR