5-Bromo-7,7-dimethyl-7H-benzo[c]fluorene 97%
Supplier: Ambeed
|
Warning
|
A141181-100MG
A141181-1G
A141181-250MG
A141181-5G
77234-986EA
22.66
USD
77234-986
77234-988
77234-990
77234-992
5-Bromo-7,7-dimethyl-7H-benzo[c]fluorene 97%
5-Bromo-7,7-dimethyl-7H-benzo[c]fluorene
Precaución: for Research Use Only
Formula:
C₁₉H₁₅Br MW: 323.23 g/mol |
MDL Number:
MFCD28975085 CAS Number: 954137-48-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...