6-Chloro-o-toluic acid 98%

Supplier: Ambeed
Warning

A141168-10G A141168-1G A141168-250MG A141168-25G A141168-5G A141168-100G
77234-976EA 29 USD
77234-976 77234-978 77234-980 77234-982 77234-984 77615-124
6-Chloro-o-toluic acid 98%
6-Chloro-o-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₂
MDL Number: MFCD00045799
CAS Number: 21327-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR