(±)-3,5-Difluoromandelic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3,5-Difluoromandelic acid, (±)-3,5-Difluorophenylglycolic acid

A141148-1G A141148-250MG A141148-5G
77234-972EA 44 USD
77234-972 77234-974 77592-676
(±)-3,5-Difluoromandelic acid 95%
(±)-3,5-Difluoromandelic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₃
MDL Number: MFCD00061300
CAS Number: 132741-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR