2,4-Bis(4-methylpiperazin-1-yl)aniline 97%

Supplier: Ambeed
Warning

A141063-100MG A141063-1G A141063-250MG
77234-966EA 48.41 USD
77234-966 77234-968 77234-970
2,4-Bis(4-methylpiperazin-1-yl)aniline 97%
2,4-Bis(4-methylpiperazin-1-yl)aniline

Precaución: for Research Use Only

Formula: C₁₆H₂₇N₅
MW: 289.41 g/mol
MDL Number: MFCD05239277
CAS Number: 797813-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR