NSC-348884 98%

Supplier: Ambeed
Warning

Synonyms: NSC348884, N1,N1,N2,N2-Tetrakis((6-methyl-1H-benzo[d]imidazol-2-yl)methyl)ethane-1,2-diamine, N1,N1,​N2,​N2-​tetrakis[(6-​methyl-​1H-​benzimidazol-2-​yl)​methyl]-1,​2-ethanediamine

A141034-10MG A141034-1MG A141034-25MG A141034-50MG A141034-5MG
77234-956EA 20 USD
77234-956 77234-958 77234-960 77234-962 77234-964
NSC-348884 98%
NSC-348884

Precaución: for Research Use Only

Formula: C₃₈H₄₀N₁₀
MDL Number: MFCD12546137
CAS Number: 81624-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR