8-Bromo-6-chloro-2-methylimidazo[1,2-b]pyridazine 95%

Supplier: Ambeed
Warning

A141011-100MG A141011-5G
77234-952EA 61 USD
77234-952 77234-954
8-Bromo-6-chloro-2-methylimidazo[1,2-b]pyridazine 95%
8-Bromo-6-chloro-2-methylimidazo[1,2-b]pyridazine

Precaución: for Research Use Only

Formula: C₇H₅BrClN₃
MDL Number: MFCD29760620
CAS Number: 1298031-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR