5-Amino-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione 97%

Supplier: Ambeed
Warning

A140942-100MG A140942-1G A140942-250MG A140942-5G A140942-25G
77234-944EA 14 USD
77234-944 77234-946 77234-948 77234-950 77745-999
5-Amino-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione 97%
5-Amino-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₁₃H₁₁N₃O₄
MDL Number: MFCD30188077
CAS Number: 191732-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR