1-(4-Nitrophenyl)-4-piperidinol 97%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Nitrophenyl)piperidin-4-ol, 4-Hydroxy-1-(4-nitrophenyl)piperidine

A140912-1G A140912-250MG A140912-5G
77285-742EA 32 USD
77285-742 77285-744 77285-746
1-(4-Nitrophenyl)-4-piperidinol 97%
1-(4-Nitrophenyl)-4-piperidinol

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O₃
MW: 222.2400 g/mol
MDL Number: MFCD00215518
CAS Number: 79421-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR