Boc-D-His(Trt)-OH 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-D-His(trt)-OH
A140904-1G
A140904-250MG
A140904-5G
A140904-100G
77258-796EA
18.54
USD
77258-796
77258-798
77258-800
77615-116
Boc-D-His(Trt)-OH 98%
Boc-D-His(Trt)-OH
Precaución: for Research Use Only
Formula:
C₃₀H₃₁N₃O₄ MW: 497.6 g/mol |
MDL Number:
MFCD08274494 CAS Number: 393568-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...