4-Methoxybenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: Anisyl alcohol, p-Anisyl Alcohol, 4-(Hydroxymethyl)anisole, Anise alcohol, (4-Methoxyphenyl)methanol

A140857-1000G A140857-100G A140857-25G A140857-500G
77285-724EA 43.26 USD
77285-724 77285-726 77285-728 77285-730
4-Methoxybenzyl alcohol 98%
4-Methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
MDL Number: MFCD00004653
CAS Number: 105-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR