2-Acetyl-5-chlorothiophene 98%

Supplier: Ambeed
Warning

A140845-100G A140845-10G A140845-25G A140845-5G
77246-926EA 83 USD
77246-926 77246-928 77246-930 77246-932
2-Acetyl-5-chlorothiophene 98%
2-Acetyl-5-chlorothiophene

Precaución: for Research Use Only

Formula: C₆H₅ClOS
MDL Number: MFCD00005444
CAS Number: 6310-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR