SHP099 hydrochloride 98+%

Supplier: Ambeed
Warning

A140769-10MG A140769-1MG A140769-250MG A140769-5MG A140769-100MG A140769-1G A140769-50MG
77234-930EA 28 USD
77234-930 77234-932 77234-934 77234-936 77745-994 77745-995 77745-996
SHP099 hydrochloride 98+%
SHP099 hydrochloride

Precaución: for Research Use Only

Formula: C₁₆H₂₀Cl₃N₅
MDL Number: MFCD30343849
CAS Number: 1801747-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR