(E/Z)-BIX02189 97%

Supplier: Ambeed
Warning

Synonyms: 3-[[[3-[(Dimethylamino)methyl]phenyl]amino]phenylmethylene]-2,3-dihydro-N,N-dimethyl-2-oxo-1H-indole-6-carboxamide, BIX-02189 (Random Configuration), BIX-02189

A140614-10MG A140614-1MG A140614-25MG A140614-50MG A140614-5MG
77258-782EA 28 USD
77258-782 77258-784 77258-786 77258-788 77258-790
(E/Z)-BIX02189 97%
(E/Z)-BIX02189

Precaución: for Research Use Only

Formula: C₂₇H₂₈N₄O₂
MDL Number: MFCD18074528
CAS Number: 1094614-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR