Methyl-3-chloro-4-hydroxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-chloro-4-hydroxybenzoate

A140601-100G A140601-10G A140601-1G A140601-250MG A140601-25G A140601-5G
77234-918EA 227.63 USD
77234-918 77234-920 77234-922 77234-924 77234-926 77234-928
Methyl-3-chloro-4-hydroxybenzoate 98%
Methyl-3-chloro-4-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
MW: 186.59 g/mol
MDL Number: MFCD00016418
CAS Number: 3964-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR