[1,1'-Biphenyl]-2,4-diol 97%

Supplier: Ambeed
Warning

A140513-100MG A140513-1G
77246-916EA 9.27 USD
77246-916 77246-918
[1,1'-Biphenyl]-2,4-diol 97%
[1,1'-Biphenyl]-2,4-diol

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂
MW: 186.20 g/mol
MDL Number: MFCD00029715
CAS Number: 134-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR