Nutlin-3 98%

Supplier: Ambeed
Warning

A140511-100MG A140511-1MG A140511-250MG A140511-5MG A140511-50MG A140511-10MG
77258-774EA 242 USD
77258-774 77258-776 77258-778 77258-780 77615-114 77747-988
Nutlin-3 98%
Nutlin-3

Precaución: for Research Use Only

Formula: C₃₀H₃₀Cl₂N₄O₄
MDL Number: MFCD07784509
CAS Number: 548472-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR