4-(1-Methyl-1H-pyrazol-3-yl)benzaldehyde 96%

Supplier: Ambeed
Warning

A140490-100MG A140490-1G A140490-250MG
77285-708EA 43.26 USD
77285-708 77285-710 77285-712
4-(1-Methyl-1H-pyrazol-3-yl)benzaldehyde 96%
4-(1-Methyl-1H-pyrazol-3-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O
MW: 186.2100 g/mol
MDL Number: MFCD08669906
CAS Number: 179055-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR