(S)-2-Amino-2-(3-(trifluoromethoxy)phenyl)acetic acid 97%

Supplier: Ambeed
Warning

A140468-100MG A140468-1G A140468-250MG A140468-5G
77246-904EA 33.99 USD
77246-904 77246-906 77246-908 77246-910
(S)-2-Amino-2-(3-(trifluoromethoxy)phenyl)acetic acid 97%
(S)-2-Amino-2-(3-(trifluoromethoxy)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₈F₃NO₃
MW: 235.15 g/mol
MDL Number: MFCD07371775
CAS Number: 1228547-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR