5-Iodo-3-nitropyridin-2-ol 97%

Supplier: Ambeed
Warning

A140461-100MG A140461-1G A140461-250MG
77258-764EA 5 USD
77258-764 77258-766 77258-768
5-Iodo-3-nitropyridin-2-ol 97%
5-Iodo-3-nitropyridin-2-ol

Precaución: for Research Use Only

Formula: C₅H₃IN₂O₃
MDL Number: MFCD09743049
CAS Number: 25391-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR