1,3,3-Trimethyl-6'-morpholinospiro[indoline-2,3'-naphtho[2,1-b][1,4]oxazine] 97%

Supplier: Ambeed
Warning

A140452-100MG A140452-1G A140452-250MG A140452-5G
77246-898EA 6.18 USD
77246-898 77246-900 77246-902 77745-992
1,3,3-Trimethyl-6'-morpholinospiro[indoline-2,3'-naphtho[2,1-b][1,4]oxazine] 97%
1,3,3-Trimethyl-6'-morpholinospiro[indoline-2,3'-naphtho[2,1-b][1,4]oxazine]

Precaución: for Research Use Only

Formula: C₂₆H₂₇N₃O₂
MW: 413.51 g/mol
MDL Number: MFCD00329492
CAS Number: 114747-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR