2,4,6-Tribromophenol 98%

Supplier: Ambeed
Danger

A140105-1000G A140105-100G A140105-25G A140105-500G
77246-890EA 44 USD
77246-890 77246-892 77246-894 77246-896
2,4,6-Tribromophenol 98%
2,4,6-Tribromophenol

Precaución: for Research Use Only

Formula: C₆H₃Br₃O
MDL Number: MFCD00002150
CAS Number: 118-79-6
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR