8-Benzyl-1-cyclohexyl-1,3,8-triazaspiro[4.5]decan-4-one 97%

Supplier: Ambeed
Warning

A139972-1G A139972-250MG
77258-748EA 41.2 USD
77258-748 77258-750
8-Benzyl-1-cyclohexyl-1,3,8-triazaspiro[4.5]decan-4-one 97%
8-Benzyl-1-cyclohexyl-1,3,8-triazaspiro[4.5]decan-4-one

Precaución: for Research Use Only

Formula: C₂₀H₂₉N₃O
MW: 327.46 g/mol
MDL Number: MFCD30729971
CAS Number: 95939-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR