6-Methoxysalicylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-6-methoxybenzoic acid, 6-Hydroxy-o-anisic acid

A139965-100G A139965-10G A139965-1G A139965-25G A139965-5G
77246-880EA 247.2 USD
77246-880 77246-882 77246-884 77246-886 77246-888
6-Methoxysalicylic acid 97%
6-Methoxysalicylic acid

Precaución: for Research Use Only

Formula: C₈H₈O₄
MW: 168.15 g/mol
MDL Number: MFCD00674090
CAS Number: 3147-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR