2-(2-Chlorophenyl)ethanol 98%

Supplier: Ambeed
Warning

A139865-10G A139865-5G
77258-744EA 46.35 USD
77258-744 77258-746
2-(2-Chlorophenyl)ethanol 98%
2-(2-Chlorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₉ClO
MW: 156.61 g/mol
MDL Number: MFCD00002888
CAS Number: 19819-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR