3,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 3,5-Dimethylpyrazole-4-boronic acid pinacol ester

A139751-100G A139751-10G A139751-1G A139751-250MG A139751-25G A139751-5G
77272-440EA 299 USD
77272-440 77272-442 77272-444 77272-446 77272-448 77272-450
3,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester 97%
3,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₉BN₂O₂
MDL Number: MFCD05663852
CAS Number: 857530-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR