Silver(I) trifluoromethanethiolate 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
Trifluoromethanethiol Silver(I) salt
A139672-1G
A139672-250MG
A139672-5G
A139672-25G
77246-866EA
44
USD
77246-866
77246-868
77549-698
77745-988
Silver(I) trifluoromethanethiolate 95%
Silver(I) trifluoromethanethiolate
Precaución: for Research Use Only
Formula:
CAgF₃S MW: 208.93 g/mol |
MDL Number:
MFCD25563234 CAS Number: 811-68-7 UN: 1759 ADR: 8 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...