Methyl-2-bromo-4-nitrobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-bromo-4-nitrobenzoate, 2-Bromobenzoic acid methyl ester

A139667-100G A139667-10G A139667-1G A139667-25G A139667-5G
77234-862EA 224.54 USD
77234-862 77234-864 77234-866 77234-868 77234-870
Methyl-2-bromo-4-nitrobenzoate 97%
Methyl-2-bromo-4-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrNO₄
MW: 260.04 g/mol
MDL Number: MFCD07779271
CAS Number: 100959-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR