4-Chloro-7-methyl-1H-indole 97%

Supplier: Ambeed
Warning

A139636-100MG A139636-1G A139636-250MG
77285-686EA 7 USD
77285-686 77285-688 77285-690
4-Chloro-7-methyl-1H-indole 97%
4-Chloro-7-methyl-1H-indole

Precaución: for Research Use Only

Formula: C₉H₈ClN
MDL Number: MFCD09027024
CAS Number: 61258-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR