Methyl 2-amino-3-chloropropanoate hydrochloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-DL-β-Cl-Ala-OMe·HCl
A139598-100MG
A139598-1G
A139598-250MG
A139598-5G
A139598-25G
77285-668EA
16.48
USD
77285-668
77285-670
77285-672
77285-674
77615-112
Methyl 2-amino-3-chloropropanoate hydrochloride 95%
Methyl 2-amino-3-chloropropanoate hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₉Cl₂NO₂ MW: 174.02 g/mol |
MDL Number:
MFCD00038914 CAS Number: 33646-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...