2,6-Difluorobenzoic acid 97%

Supplier: Ambeed
Warning

A139538-10G A139538-25G A139538-5G A139538-500G A139538-100G
77272-434EA 6 USD
77272-434 77272-436 77272-438 77566-796 77615-110
2,6-Difluorobenzoic acid 97%
2,6-Difluorobenzoic acid

Precaución: For research use only.

Formula: C₇H₄F₂O₂
MDL Number: MFCD00002411
CAS Number: 385-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR