4-(Trifluoromethoxy)benzylamine 98%

Supplier: Ambeed
Warning

Synonyms: (4-(Trifluoromethoxy)phenyl)methanamine

A139508-10G A139508-1G A139508-25G A139508-5G
77246-850EA 13 USD
77246-850 77246-852 77246-854 77246-856
4-(Trifluoromethoxy)benzylamine 98%
4-(Trifluoromethoxy)benzylamine

Precaución: for Research Use Only

Formula: C₈H₈F₃NO
MDL Number: MFCD00061237
CAS Number: 93919-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR