2-Fluoro-4-nitrobenzoic acid 97%

Supplier: Ambeed
Warning

A139445-100G A139445-10G A139445-1G A139445-250MG A139445-25G A139445-500G A139445-5G
77272-420EA 74 USD
77272-420 77272-422 77272-424 77272-426 77272-428 77272-430 77272-432
2-Fluoro-4-nitrobenzoic acid 97%
2-Fluoro-4-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄FNO₄
MDL Number: MFCD00275565
CAS Number: 403-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR