tert-Butyl 2-chloropyrimidine-4-carboxylate 97%

Supplier: Ambeed
Warning

A139311-100MG A139311-250MG A139311-5G
77234-856EA 38 USD
77234-856 77234-858 77234-860
tert-Butyl 2-chloropyrimidine-4-carboxylate 97%
tert-Butyl 2-chloropyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₁ClN₂O₂
MDL Number: MFCD29760807
CAS Number: 220041-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR