p-Tolyldiethanolamine 95%

Supplier: Ambeed
Danger

Synonyms: N,N-Bis(2-hydroxyethyl)-p-toluidine, 2,2'-(4-Methylphenylimino)diethanol, 2,2'-(p-Tolylimino)diethanol, 2,2'-(p-Tolylazanediyl)diethanol

A139257-1000G A139257-100G A139257-10G A139257-25G A139257-500G
77246-836EA 70.04 USD
77246-836 77246-838 77246-840 77246-842 77246-844
p-Tolyldiethanolamine 95%
p-Tolyldiethanolamine

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₂
MW: 195.26 g/mol
MDL Number: MFCD00020580
CAS Number: 3077-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR