5-Hydroxy-3H-benzimidazole 98%

Supplier: Ambeed
Warning

A139233-1G A139233-250MG A139233-25G A139233-5G
77234-848EA 39 USD
77234-848 77234-850 77234-852 77234-854
5-Hydroxy-3H-benzimidazole 98%
5-Hydroxy-3H-benzimidazole

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MDL Number: MFCD02241303
CAS Number: 41292-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR