2-(4-(1-(Quinolin-6-ylmethyl)-1H-[1,2,3]triazolo[4,5-b]pyrazin-6-yl)-1H-pyrazol-1-yl)ethan-1-ol methanesulfonate 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
PF-04217903 mesylate
A139192-10MG
A139192-1MG
A139192-50MG
A139192-5MG
77246-828EA
42
USD
77246-828
77246-830
77246-832
77246-834
2-(4-(1-(Quinolin-6-ylmethyl)-1H-[1,2,3]triazolo[4,5-b]pyrazin-6-yl)-1H-pyrazol-1-yl)ethan-1-ol methanesulfonate 98+%
2-(4-(1-(Quinolin-6-ylmethyl)-1H-[1,2,3]triazolo[4,5-b]pyrazin-6-yl)-1H-pyrazol-1-yl)ethan-1-ol methanesulfonate
Precaución: for Research Use Only
Formula:
C₂₀H₂₀N₈O₄S MW: 468.49 g/mol |
MDL Number:
MFCD17392584 CAS Number: 956906-93-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...